missing translation for 'onlineSavingsMsg'
Learn More

Idex Fingertight Nut, 1/8 in., 1/4-28 Coned; Kel-F (PCTFE)

Catalog No. 0570067
Encompass
Change view
Click to view available options
Material:
PCTFE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
0570067 PCTFE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0570067 Supplier Idex Supplier No. UP P 100
Only null left

Specifications

Type Diamond Knurl Male Nut Fitting; 0.25 to 28 Coned
Material PCTFE
Color Natural
For Use With (Equipment) For 0.12 in. O.D. Tubing