Learn More
Idex Peristaltic Tubing Adapters
Catalog No. 05700193
Encompass
Change view
Click to view available options
Diameter (English) Inner:
0.048 to 0.110 in.
Catalog No. Diameter (English) Inner
05700193 0.048 to 0.110 in.
1 options
Catalog No. 05700193 Supplier Idex Supplier No. UP P 757/DUP
Connect 1/16 in. OD rigid/semi-rigid tubing to soft-wall tubing
Peristaltic tubing adapters feature specially designed nose to allow soft tubing to fit over the nose then be held in place by the retainer sleeve. To connect peristaltic tubing to rigid tubing with a different OD, order a flangeless fitting with 1/4-28 flat-bottom configuration.
Specifications
Type Peristaltic Tubing Adapter
Diameter (English) Inner 0.048 to 0.110 in.
For Use With (Equipment) Peristaltic to 1/16 in. O.D. Rigid