missing translation for 'onlineSavingsMsg'
Learn More

Idex Fingertight Ferrules

Catalog No. 057007
Encompass
Change view
Click to view available options
For Use With (Equipment):
0.0625 in. O.D. Tubing
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
057007 0.0625 in. O.D. Tubing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 057007 Supplier Idex Supplier No. UP F 142X

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

For Use With (Equipment) 0.0625 in. O.D. Tubing
Material PEEK
Type Fingertight Ferrule
Color Natural