Learn More
Idex Barbed-to-Threaded Adapters
Catalog No. 29008412
Encompass
Change view
Click to view available options
Diameter (Metric) Inner:
1.57 mm
4.76 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
29008412 1.57 mm
290081375 4.76 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 29008412 Supplier Idex Supplier No. UP P 646
Only null left
Easily connect soft-wall tubing to 10-32 coned, 1/4-28 flat-bottom, or 5/16-24 flat-bottom port
Slip soft-wall tubing over the barbed end then thread the adapter into a receiving port for a reliable low-pressure connection.
Specifications
Type Adapter
Diameter (Metric) Inner 1.57 mm
For Use With (Application) Connecting Soft Tubing to Standard 1/4-28 Internally-Threaded Ports
Material ETFE