Learn More
Idex Adapters for HPLC
Convenience of quick connect and disconnect of a luer connection
$10.17 - $54.47
Products 4
Catalog Number Mfr. No. Connections Material Price Quantity & Availability  
Catalog Number Mfr. No. Connections Material Price Quantity & Availability  
05700141
Encompass
View Documents Promotion Details
Idex
UP P 604
Female Luer to 1/4-28 Flat Bottom Acetal
Each for $10.17
Only null left
05700154
Encompass
View Documents Promotion Details
Idex
UP P625
Male Luer to 1/4-28 Flat Bottom ETFE
Each for $13.45
Only null left
05700167
Encompass
View Documents Promotion Details
Idex
UP P 642/DUP
Female Luer to Male 10-32 Adapter ETFE
Each for $13.74
Only null left
0570074
Encompass
View Documents Promotion Details
Idex
UPP 135
1/4-28 Flat Bottom Female to 5/16-24 Female Flat Bottom PEEK
Each for $54.47
Only null left
Description
Description
Convenience of quick connect and disconnect of a luer connection
  • Manufactured from inert polymers
Quick connect luer adapters are designed to connect any female luer to any male luer in a variety of applications. Use with male luer lock adapters for more secure connection.