Learn More
MilliporeSigma™ Supelco™ Upchurch In-Line Cartridge Check Valve
Description
The cartridge and flangeless fitting is for use with 1/16 in. O.D. tubing. The internal volume of the check valve is less than 150 μL. Place in the flow system where you wish to restrict fluid flow to one direction. These assemblies withstand system pressures of 1.000 psi (69 bar). Complete with wrench.
Specifications
Specifications
| For Use With (Application) | Chromatography Applications |