missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals uPA Inhibitor II, UK122

Catalog No. AAJ64874LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64874LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64874LB0 Supplier Thermo Scientific Chemicals Supplier No. J64874LB0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

C17H13N3O2·CF3CO2H

A cell-permeable oxazolidinone-carboxamidine compound that acts as a potent, selective, and active-site targeting inhibitor of urokinase-type plasminogen activatoruPA Inhibitor II, UK122 acts as a potent, selective and active-site targeting urokinase-type plasminogen activator (uPA) inhibitor. However, its activity against trypsin, thrombin, tPA, and plasmin is greatly reduced. It is also effectively inhibits the migration (80% inhibition) and invasion (68% inhibition) of uPA-expressing CFPAC-1 pancreatic cells, without affecting its proliferation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A cell-permeable oxazolidinone-carboxamidine compound that acts as a potent, selective, and active-site targeting inhibitor of urokinase-type plasminogen activator

Specifications

Quantity 5 mg
Description Yellow
Content And Storage 2°C to 8°C
For Use With (Application) A cell-permeable oxazolidinone-carboxamidine compound that acts as a potent, selective, and active-site targeting inhibitor of urokinase-type plasminogen activator
Molecular Formula C17H13N3O2·CF3CO2H
Formula Weight 405.30g/mol
Sensitivity Light Sensitive
Product Type Signal Transduction Reagent
Form Solid

RUO – Research Use Only