missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ uPA Activity Assay Kit

Catalog No. ECM600
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ECM600 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ECM600 Supplier MilliporeSigma™ Supplier No. ECM600

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • uPA Positive Control: One lyophilized vial, 1000 units, of uPA from human urine
  • Chromogenic Substrate: One 5mg bottle of Tripeptide with pNA group
  • Assay Buffer, 10X: One 5mL bottle

Provides a quick, efficient and sensitive system for evaluation of uPA activity and for screening of uPA inhibitors

  • The assay is colorimetric and does not require radioactivity or fluorescence equipment
  • The assay is sensitive over a range of 0.05-50 units of uPA activity
  • Utilizes a chromogenic substrate, which is cleaved by active uPA
  • Addition of this substrate to a uPA-containing sample results in a colored product, detectable by its Optical Density at 405nm (OD405) 

Specifications

Content And Storage Store kit materials at -20°C for up to 6 months
Detection Method Colorimetric
For Use With (Application) Activity Assay
Quantity 1 Kit