missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Unlined Pigskin Leather Drivers Gloves

Catalog No. p-145136
Change view
Click to view available options
Size:
Small
Medium
X-Large
Catalog No. Size
19043035 Small
19043030 Medium
19043042 X-Large
3 options

Catalog No. 19043035

Supplier: Fisherbrand™ 70301/S

Only null left

Gloves return to their original pliability even after being soaked in water.

  • Suitable for construction, machine operation, utility work and many general applications
  • Gunn-cut style with straight thumb
  • Elastic wrist provides a sure fit
  • Unlined

Specifications

Color Natural
Material Pigskin Leather
Product Type Driver Gloves
Size Small
Style Gunn
For Use With (Application) Construction, Machine Operation, Utility Work and General Applications
Glove Cuff Style Open
Hem Color Red
Thumb Style Straight
Length (English) 9 in.
Length (Metric) 22.8 cm
Show More Show Less