missing translation for 'onlineSavingsMsg'
Learn More

Basix™ Universal Pipette Tips

The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.

 View more versions of this product

Catalog No. 13-611-112



Only null left

Description

Description

The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.

Basix offers reliability and quality for all varieties of experiments, from everday liquid transfer to PCR and Electrophoresis, Basix offers a tip to meet your needs.

Choose from non-Sterile or Sterile with packaging options including bulk, racked, reloads, stack racks.

  • Soft pliable tip collar allows this tip to fit on pipets made by various manufacturers
  • Lower cost and increased convenience by purchasing fewer products
  • Available in a variety of packaging options
  • Eliminate the need to buy pipetter-specific tips
  • Certified by independent laboratory
  • Sterile: RNase, DNase, DNA and Pyrogen free
  • Available in Natural or Blue (13611123 and 13611214)
  • Nonsterile: RNase, DNase and DNA free
Specifications

Specifications

200 μL
Natural
2 in.
DNase, RNase, DNA, Pyrogen Free
Universal
Polypropylene
Non-sterile
Autoclavable
5.08 cm
Stack Rack
Universal Fit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions