missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Universal Sleeve Protectors

Description
- Disposable general-purpose sleeves
- Latex free
- Ultrasonic sewing
- Handmade
- Dual-end elastic allows for a secure fit
Specifications
Specifications
| Product Type | Controlled Environment Sleeve |
| Size | Universal |
| Construction | Sleeve Cover |
| Material | Microporous Fabric |
| Color | White |
| Sterility | Non-sterile |
| Disposable | Single-use |
| Closure Type | Elastic Ends |
| Seam Type | Ultrasonically Welded |
| Cuff Style | Elastic |
| Show More |