Learn More
Restek™ Universal Press-Tight™ Connectors: Universal Y
Catalog No. 06710612
Encompass
Change view
Click to view available options
Includes:
Undeactivated
Quantity:
1 Ea.
3 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Quantity
06710612 Undeactivated 1 Ea.
06717754 Undeactivated 3 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06710612 Supplier Restek™ Supplier No. RK 20405
Connect analytical columns, guard columns, transfer lines or detectors
  • Fit column outer diameters 0.33 to 0.74mm (Restek inside diameters 0.1 to 0.53mm)
  • Deactivated Press-Tight connectors for better recovery of polar and nonpolar compounds
  • Angle approximates the curvature of a capillary column, reducing the strain on column-end connections
Specifications
Type Universal Y Press-Tight Connector
Includes Undeactivated
Quantity 1 Ea.