missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 10000 Training Suit

Catalog No. 18888880
Change view
Click to view available options
Size:
Medium
3X-Large
4X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18888880 Medium
18888878 3X-Large
18888879 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18888880 Supplier DuPont™ Supplier No. TK586SLYMD000100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Offers a high level of protection against toxic, corrosive gases and liquid chemicals

  • Tychem™ 10000 fully encapsulated training suits offer an economical and effective option for hazmat training
    • Same fabric and garment design as standard Tychem™ 10000 suits
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Extra wide (EX) single layer face shield (20 mil PVC) for wide viewing angle
  • Front entry design allows wearer to partially don garment (maintain standby position) without assistance
  • Integrated butyl gloves
  • Internal adjustable waist belt system for support and improved fit
  • Clearly labeled for training use only
  • Manufactured under specifications that do not contain natural rubber latex
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant

Specifications

Color Lime Yellow
For Use With (Application) Hazmat and Emergency Response Training
Material Tychem
Size Medium
Ankle Style Boot Overflaps
Closure Type Zipper
Product Type Encapsulated Training Suit
Seam Type Serged