missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Universal Mouse Reference RNA for QuantiGene Assays

Catalog No. QS0640
Encompass
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QS0640 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. QS0640 Supplier Invitrogen™ Supplier No. QS0640

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Universal Mouse Reference RNA is an RNA extract used as a positive control in QuantiGene assays.

Universal Mouse Reference RNA is an RNA extract used as a positive control in QuantiGene assays. The Universal Mouse Reference RNA is composed of total RNA from 11 mouse cell lines:

  • Embryo
  • Embryo, fibroblast
  • Kidney
  • Liver, hepatocyte
  • Lung, alveolar macrophage
  • B-lymphocyte
  • T-lymphocyte (thymus)
  • Mammary gland
  • Muscle myoblast
  • Skin
  • Testis

Equal quantities of DNase-treated total RNA from each cell line were pooled to make the universal control.

Specifications

Species Mouse
Sample Type RNA
For Use With (Application) Nucleic Acid Quantitation
Product Type Reference RNA
Quantity 10 μg
Sample Source Cell Line
Shipping Condition Shipped at 2°C to 8°C

For Research Use Only. Not for use in diagnostic procedures.