missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Universal Linear Pipette Stand

Description
Axygen™ universal linear stand for pipettors, (single-channel, 8-channel or 12-channel pipettors)
Specifications
Specifications
| Color | Clear |
| Holds | Single-channel or Multi-channel Pipettor (1) |
| Positions | 1 |
| For Use With (Application) | Liquid Handling |
| For Use With (Equipment) | Pipettes |
| Product Type | Pipette Stand |