missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Universal Indicator, Bogens, Alcoholic, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Universal Indicator, Bogens |
| Name Note | Alcoholic |
| CAS | 67-63-0, 7732-18-5, 62625-21-2, 76-59-5, 60-11-7, 493-52-7 |
| CAS Min % | 63.0, 34.57, 0.06, 0.05, 0.04, 0.03 |
| CAS Max % | 66.0, 35.98, 0.07, 0.05, 0.04, 0.03 |
| Packaging | Amber Glass Bottle |
| Color | Orange/Red |
| Quantity | 120 mL |
| Physical Form | Liquid |
| Grade | Indicator |