Learn More
Eisco™ Universal Clamp with Holder
Catalog No. S24282
Change view
Click to view available options
Color:
Silver
Catalog No. Color
S24282 Silver
1 options
Catalog No. S24282 Supplier Eisco™ Supplier No. CH0688B
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This universal clamp is manufactured with stamped steel. One side of the jaw is flat while the other side is a 'V' shape. Designed for cylindrical shaped objects.
Specifications
Type Clamp
Material Steel
Color Silver
Jaw Type Flat Prong