Learn More
Thermo Scientific™ Unity Lab Services™ UV-Vis Instrument Installation Plans
Catalog No. 701674400
Change view
Click to view available options
Product Type:
Basic Installation Plan
Validated Installation Plan
For Use With (Equipment):
Evolution 200 UV/Vis Spectrophotometer
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment)
701674400 Validated Installation Plan Evolution 200 UV/Vis Spectrophotometer
701674300 Basic Installation Plan Evolution 200 UV/Vis Spectrophotometer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 701674400 Supplier Thermo Scientific™ Supplier No. 701674400
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Installation options for your Thermo Scientific UV-Vis Spectrophotometer
Installation
  • Includes on-site installation of instrument
  • Includes basic instrument training
  • Does not include instrument performance check or Qualification service
Validated Installation
  • Includes on-site installation of instrument
  • Includes installation qualification and operational qualification
  • Includes basic instrument / software training
  • Requires separately purchased standards for ongoing instrument qualification
  • Requires separate purchase of UV Validator
Specifications
For Use With (Equipment) Evolution 200 UV/Vis Spectrophotometer
Product Type Validated Installation Plan
Service Offering On-site installation of instrument, installation qualification and operational qualification, basic instrument/software training
For Use With (Application) On-site installation options for your UV-Vis Spectrophotometer