missing translation for 'onlineSavingsMsg'
Learn More

United Scientific Supplies Wheel and Axle

Catalog No. S13049
Change view
Click to view available options
Grade:
K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S13049 K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13049 Supplier United Scientific Supplies Supplier No. WAXL01

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For demonstrating mechanical advantage and conservation of energy.

  • Consists of single stepped wheel with four coaxial steps of 35, 50, 65, and 105mm diameter
  • Wheel is supported on steel rod by ball bearing

Specifications

Grade K to 12
Format Model
Product Type Wheel and axle
Languages English
Subject Physics