Learn More
United Scientific Supplies United Scientific™ Turbidity Tube
Description
The clear acrylic turbidity tube features a small Secchi disk at the bottom and a metric scale printed along the side for easy readings. To determine turbidity, fill the tube to the top, then slowly release water using the drainage hose until the Secchi disk becomes visible when viewed from above. Clamp the hose, hold the tube level, and record the water level at the scale. Repeat for additional water samples, working efficiently so suspended particles do not settle. Higher water levels indicate clearer water, while lower water levels indicate higher turbidity.
- Clear acrylic tube with integral Secchi disk and metric scale
- Easy-to-read graduations for repeatable field or lab measurements
- One-hand operable hose clamp for controlled water release
- Compact length supports field portability
- Includes access to a downloadable activity guide
Specifications
Specifications
| Type | Turbidity Tube |
| Includes | Downloadable Activity Guide |
| Material | Acrylic |
| For Use With (Application) | Direct Turbidity Readings |