Learn More
United Scientific Supplies Plain Wire Gauze
Plain Wire Gauze used for general heating purposes in laboratories.
Supplier:  United Scientific Supplies WGPL66PK/12
 View more versions of this product
Catalog No. S01295
Only null left
Description
Description
  • Iron wire gauze (24 SWG/10 Mesh) with folded edges
  • Sold as packs of 10 or 12.
  • Available in three different sizes
Specifications
Chemical Identifiers
No Ceramic Center
Iron
Folded
Square
Specifications
Folded
Square
Iron
6 in.
15.2 cm
15.2 cm
6 in.
No Ceramic Center
Wire Mesh
15.2 x 15.2 cm (6 x 6 in.)
12 Pk.
Product Suggestions
Product Suggestions