missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ Graduated Polypropylene Pitchers with Handle
These polypropylene beakers include a handle and easy to read printed graduations.
$204.21 - $204.21
Specifications
| Material | Polypropylene |
|---|---|
| Includes | Spout |
| Type | Pitcher |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
Description
High Quality polypropylene beakers.
- Tapered design is convenient for stacking and gripping
- Beakers include pour spout and easy-to-read silk-screened graduations
- Features a comfortable rectangular handle
- Autoclavable
- Available in sizes ranging from 100 to 10,000 mL
Specifications
| Polypropylene | |
| Pitcher | |
| Reusable | |
| Silk Screened Print Graduated |
| Spout | |
| Autoclavable | |
| Yes | |
| Closed Handle |