missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific Supplies United Scientific™ Sun Tracking Kit
Description
Includes 10 polystyrene hemisphere domes, a compass, wax pencil, and activity guide. Accommodates groups of up to 10 students and supports single-day observations or extended multi-month studies. Repeated seasonal observations help students compile data showing how solar declination changes throughout the year, connecting classroom theory to observable astronomical phenomena.
- Ideal for astronomy and earth science curriculum integration
- Complete 10-student kit with hemisphere domes and tracking tools
- Demonstrates seasonal solar patterns and Earth axial tilt effects
- Washable hemispheres can be used multiple times
- Includes downloadable activity guide
Specifications
Specifications
| Grade Level | Intermediate - Middle School, High School, Higher Ed |
| Grade | 5 to 12 |
| Format | Kit |
| Product Type | Sun Tracking Kit |
| Languages | English |
| Subject | Earth Science |