Learn More
United Scientific Supplies United Scientific™ Spark Timer Discs
Description
Spark Timer is used for measuring velocity and acceleration of falling or moving objects. When tape is pulled through the timer, a spark burns tiny dots in the tape at precise intervals. The faster the tape is pulled, the greater the distance between the dots, allowing students to calculate acceleration.
- Measures velocity and acceleration of falling or moving objects
- Creates spark marks on tape at precise intervals
- Dot spacing helps students calculate acceleration
- Includes spark tape, carbon disks, weight, and clamp
- Includes activity guide
Specifications
Specifications
| Color | Black |
| Format | Continuous |
| Product Type | Replacement Disc for Spark Timer |