missing translation for 'onlineSavingsMsg'
Learn More

United Scientific Supplies United Scientific™ Rake Retractor

Catalog No. S001343
Change view
Click to view available options
Length (English):
6.25 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
S001343 6.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001343 Supplier United Scientific Supplies Supplier No. RRET06

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rake retractor helps hold tissue or specimens in place.

  • Rake retractor constructed from stainless steel
  • For dissections
  • Stainless steel
  • Length: 158.75 mm

Specifications

Autoclavable Autoclavable
Disposable Reusable
Length (English) 6.25 in.
Length (Metric) 158.75 mm
Material Stainless Steel
Sterility Non-sterile
Product Type Rake Retractor