Learn More
United Scientific Supplies United Scientific™ Knife Switch, Screw Posts
Description
Knife Switch with screw posts for low voltage experiments (less than 36 V).
- Plastic bases.
- Available with either thumbscrew or spring binding posts.
- Available in single/double pole, single/double throw combinations
Specifications
Specifications
| Product Type | Knife Switch |
| Material | Plastic |
| For Use With (Equipment) | Double Pole, Double Throw |