missing translation for 'onlineSavingsMsg'
Learn More

United Scientific Supplies United Scientific™ Clinometer

Catalog No. S001386
Change view
Click to view available options
Format:
Class Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
S001386 Class Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Prop 65
Catalog No. S001386 Supplier United Scientific Supplies Supplier No. CLNMTR

missing translation for 'validMainframeMsg'

Prop 65

Classroom tool used to support outdoor measurement activities.

  • Easily determine the height of an object by using angles
  • This clinometer has a spring-loaded trigger that holds a pivoting arm stationary to indicate the angle above or below the horizon
  • The maximum angle above or below the horizon is 70°

Specifications

Grade Level Intermediate - Middle School, High School
Format Class Set
Product Type Clinometer
Languages English
Subject Earth Science