missing translation for 'onlineSavingsMsg'
Learn More

United Scientific Supplies United Scientific™ Air Puck

Catalog No. S001402
Change view
Click to view available options
Product Type:
Air Puck
Air Puck Mini
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S001402 Air Puck Mini
S001387 Air Puck
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S001402 Supplier United Scientific Supplies Supplier No. AIRPUCKMINI

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Air pucks allow students to explore motion collisions and Newtonian mechanics with a hover puck that glides on a cushion of air.

  • Hover puck floats on a cushion of air across hard, flat surfaces for hands-on physics demonstrations.
  • Soft foam bumper helps protect the puck and surroundings, while a built-in battery-powered fan generates airflow for smooth travel over tile, wood, or linoleum.
  • Bright LED lights support engaging demonstrations of velocity, acceleration, momentum, and collision.
  • Available in 7 in. size requiring 3 AA batteries and 4 in. size requiring 3 AAA batteries. Batteries are not included. No activity guide is included.
  • Floats on a cushion of air across hard, flat surfaces
  • Soft foam bumper allows gentle rebounds off barriers
  • Available in 7 in. and 4 in. sizes

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Format Model
Product Type Air Puck Mini
Languages English
Subject Physics

Batteries not included. Not intended for carpet or rough surfaces. No activity guide is included.