missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Unistore™ Sterile Sealed Caps for Erlenmeyer Flasks

Catalog No. p-200071386
Change view
Click to view available options
Closure Size:
38 to 430 mm
45 to 430 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
S02068 45 to 430 mm
S02069 38 to 430 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S02068 Supplier United Scientific™ Supplier No. UNERFLSCAP45430
Only null left

Sealed Caps for use with sterile vented erlenmeyer flasks.

Caps are designed for sterile PC or PETG Erlenemeyer Flasks

  • Caps are made from HDPE.
  • For use with United Scientific Unistore PC or PETG Sterile Erlenmeyer Flasks
  • Available in two sizes
  • Caps are sealed

Specifications

Material HDPE
Product Type Sealed Cap
Autoclavable Not Autoclavable
Disposable Reusable
Sterility Sterile
Closure Type Screw Cap
Diameter (Metric) Outer 49.7 mm
Closure Size 45 to 430 mm