missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Unistore™ Sterile Erlenmeyer Flask Vented, PC

Flask ideal for mixing, containing, and storing solutions.

Supplier:  United Scientific™ UNERFLSPC1000

 View more versions of this product

Catalog No. S02067


Only null left
Explore available promotions

Description

Description

These sterile PC (polycarbonate) Erlenmeyer flasks are used for mixing, containing, and storing solutions. PC flasks are crystal clear and have easy to read graduation marks. These are shatterproof and are an ideal alternative to glass. Temperature range is -80 to 130°C (-112 to 266°F). These come with a vented cap: 0.20 μm hydrophobic PTFE membrane for continuous gas exchange. Cap material is HDPE. Sealed caps are also available.

  • Temperature range is -80 to 130°C (-112 to 266°F)
  • DNase/RNase free and nonpyrogenic
  • Sterilized by gamma irradiation
  • Leak proof tested
  • Produced in 100K clean room environment
Specifications

Specifications

1000 mL
Erlenmeyer Flask
Single-use
Threaded
Flat
Polycarbonate
Autoclavable
Yes
Sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions