Learn More
Red Kap™ Unisex White Specialized Cuffed Lab Coats
Catalog No. 19080344
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19080344 X-Large
19080340 X-Small
19080341 Small
19080342 Medium
19080343 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19080344 Supplier Red Kap™ Supplier No. KP70WHRGXL
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Unisex poly cotton specialized lab coat with exterior pockets and knit cuffs features a 6 gripper secure closure.
  • Chest pocket and two lower pockets for convenient storage
  • Knit cuffs that keep sleeves from interfering with work
  • Extended back coverage
  • Primary closure is snap, gripper
  • Wash care is industrial laundry for heavy soil and home wash for light soil
  • Fabric is 5 oz. performance blend poplin
  • 80% Polyester, 20% Combed Cotton
  • Wrinkle-resistant finish
  • Six gripper closure
  • One-piece, lined, notched lapel collar
  • Knit wristlet cuffs
  • Requires minimum ironing
  • No side vent openings
  • Unisex, meant to be worn over clothes
Specifications
Product Type General Purpose Lab Coat
Size X-Large
Gender Unisex
Material Polyester, Cotton
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Wrinkles
Style Standard Length
No. of Pockets 3
Autoclavable Not Autoclavable
Show More Show Less