Learn More
Simport™ Scientific Unisette™ Tissue Processing/Embedding Cassettes in Quickload™ Stacks
Description
- Keeps specimens safely submerged in liquid and are resistant to the chemical action of most histological solvents.
- Efficient flow-through slots maximize fluid exchange and ensure proper drainage.
- One-piece construction with snap-latch and hinge-lock design allows for simple one-hand operation and prevents early separation of the base and lid.
- Can be opened and closed as often as necessary and they re-lock securely without the danger of specimen loss.
- Convenient Quickload™ stacks are suitable for automated printers.
Specifications
Specifications
| Quantity | 1000/Cs. |
| Writing Face Angle | 45° |
| Color | Gray |
| Material | Acetal |
| Product Type | Cassette |