missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Union bore 0.020 in., thread 10-32

Catalog No. 111011617
Change view
Click to view available options
Union bore 0.020 in., thread 10-32
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111011617 Union bore 0.020 in., thread 10-32
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111011617 Supplier MilliporeSigma™ Supelco™ Supplier No. 57659

The PEEK zero dead volume unions (bore 0.020 in., thread 10-32) need to be operated at 5000 psi (352 kg/cm2). They include Fingertight III nuts and ferrules.

Specifications

For Use With (Application) Chromatography Applications
Description bore 0.020 in., thread 10-32