Learn More
Cytiva™ Union 1/16 in. Female to 1/16 in. Female Chromatography Fitting
Catalog No. 361041174
Change view
Click to view available options
Packaging Quantity:
5 Fittings
Catalog No. Packaging Quantity
361041174 5 Fittings
1 options
Catalog No. 361041174 Supplier Cytiva™ Supplier No. 11000339
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Works with connectors with an outer diameter of 1/16 in. For ÄKTA™ systems.
  • Use with ÄKTA start, ÄKTA go, ÄKTA pure, and ÄKTA avant
  • Use with fingertight connector 1/16 in. male; fingertight connector 1/16 in. male, narrow
Specifications
Type Fitting
Color Gray
Packaging Quantity 5 Fittings
Thread Size 1/16 in. (f)