missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Ungraduated Dropping Funnel

Features a 100 mL capacity, Rotaflow stopcock, and is fitted with a 19/26 interchangeable plastic stopper

Supplier:  Eisco™ CH0469N01

Numéro de catalogue. S27072


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

  • Cylindrical dropping funnel
  • Made of high quality, heavy duty Borosilicate 3.3 glass
  • Fitted with an interchangeable 19/26 plastic stopper
  • Stem with 19/26 cone
  • Rotaflow stopcock
  • Capacity: 100 mL
  • Ungraduated
Spécifications

Spécifications

100 mL
273.05 mm
19/26
No
1
Plastic
Dropping Funnel
Borosilicate Glass (3.3)
Clear
Addition Funnels
Interchangable Plastic Stopper
Rotaflow
19/26
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions