missing translation for 'onlineSavingsMsg'
Learn More

SP Scientific Hotpack Undercounter Glassware Washer

Catalog No. 15365210
Change view
Click to view available options
Depth (English):
22.88 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Depth (English)
15365210 22.88 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15365210 Supplier SP Scientific Supplier No. UCWHA10001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for ease of operation and flexible loading, SP Hotpack Undercounter Laboratory Glassware Washers fit under standard laboratory benches for a seamless room design. Equipped with premium features including a soft touch controller, enhanced drying, and quiet run cycles, they are engineered for durability and energy efficient operation. They provide reliable day-in, day-out performance so your laboratory glass is always ready-to-go.

  • All models come with removable top and bottom stainless-steel open racks to accommodate a variety of glassware
  • Spindle rack ready models accommodate direct injection spindle racks (sold separately) for thorough cleaning inside narrow-neck glassware
  • Stainless steel baskets, insert racks and accessories sold separately to maximize space, improve efficiency, and protect specialty glassware
  • Temperature selection to 70 ̊C (158 ̊F); can effectively dissolve stubborn oils
  • DI rinse models ensure a spotless finish; easily connect to fixed or portable DI water systems
  • Optional DI Booster Pump recommended when the incoming DI water source has a pressure of less than 18psi
  • Delayed start feature enables washer to operate unattended during off-peak hours to reduce energy costs
  • Designed for optimal quiet operation with noise buffering insulation
  • Three filters protect pumps from debris; conveniently located for easy access
  • Minimal, efficient water consumption
  • Enhanced dual fan drying system for spotless results
  • 304 stainless steel chamber ensures corrosion protection and long life
  • Adjustable leveling legs
  • UL 61010-1 & CSA 22.2 61010-1 certified by MET Labs
  • Microprocessor control system features 6 cycle selections including a customizable timed option to ensure minimum energy consumption. All wash cycles have pre-wash, wash, pre-rinse and rinse cycles to prevent contamination.
  • Rinse Only - Short tap water rinse cycle
  • Normal Wash - Lower temperature (55 ̊C) longer cycle with three (3) final rinse cycles
  • Daily Wash - Standard temperature (65 ̊C) with two (2) final rinse cycles
  • Heavy Wash - Higher temperature (70 ̊C) longer cycle with multiple pre-wash and final rinse cycles
  • Heavy Wash Time Saver - Lower temperature (55 ̊C) version of Heavy Wash with a shorter overall cycle time
  • Timed Wash Programs - Standard temperature (65 ̊C) in programmable 15 minute increments up to 2.5 hours
  • Standard warranty: 1 year

Warranty

1 year

Specifications

Description Fits standard height laboratory benches; min. height 87.5 cm (34.44 in.)
Display Digital
Material Stainless Steel
Product Type DI Rinse - Top and Bottom Standard Racks
Max. Pressure 60 psi
Water Consumption Min: Tap 2.4 Gal (9L), DI 0.8 Gal (3L); Max: Tap 4 Gal (15L), DI 1.6 Gal (6L)
Style Not compatible with a spindle rack
Certifications/Compliance MET Labs
For Use With (Application) Washing Lab. Glassware
Temperature Setting 55°-70°C (131°-158°F)
Depth (English) 22.88 in.
Depth (Metric) 58.0 cm
Height (English) Adjustable 34.44 to 36.38 in.
Height (Metric) Adjustable 87.5 to 92.4 cm
Width (English) 24.0 in.
Width (Metric) 61.0 cm
Weight (English) Shipping 116.8 lb.
Weight (Metric) Shipping 53 kg
Electrical Requirements 120V, 60Hz, 15A
Voltage 120 V
Frequency 60 Hz
Show More Show Less