missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Undecylenoyl phenylalanine (Sepiwhite) Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 175357-18-3 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Synonym | Undecylonyl Phenylalanine |
| Type | Certified Reference Material |
| Quantity | 1 g |
| Grade | Certified Reference Material |