missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Undecylenic aldehyde, 97%
CAS: 112-45-8 | C11H20O | 168.28 g/mol
$95.18 - $95.18
Chemical Identifiers
| CAS | 112-45-8 |
|---|---|
| Molecular Formula | C11H20O |
| Molecular Weight (g/mol) | 168.28 |
| MDL Number | MFCD00007032 |
| InChI Key | OFHHDSQXFXLTKC-UHFFFAOYSA-N |
| Synonym | 10-undecenal, undecylenic aldehyde, undecylenaldehyde, hendecenal, 10-undecen-1-al, 1-undecen-10-al, 10-undecylenealdehyde, 10-hendecenal, 10-undecenyl aldehyde, c-11 aldehyde, undecylenic |
| PubChem CID | 8187 |
| IUPAC Name | undec-10-enal |
| SMILES | C=CCCCCCCCCC=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC157990250
|
Thermo Scientific Chemicals
157990250 |
25 g | Glass bottle |
Each for $95.18
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 112-45-8 | |
| 168.28 | |
| OFHHDSQXFXLTKC-UHFFFAOYSA-N | |
| 8187 | |
| C=CCCCCCCCCC=O |
| C11H20O | |
| MFCD00007032 | |
| 10-undecenal, undecylenic aldehyde, undecylenaldehyde, hendecenal, 10-undecen-1-al, 1-undecen-10-al, 10-undecylenealdehyde, 10-hendecenal, 10-undecenyl aldehyde, c-11 aldehyde, undecylenic | |
| undec-10-enal |
Specifications
| 112-45-8 | |
| White | |
| 235°C | |
| Authentic | |
| Glass bottle | |
| 1.4417 to 1.4437 | |
| MFCD00007032 | |
| 0.845 | |
| Solubility in water: insoluble. Other solubilities: soluble in 80% alcohol,soluble in fixed oils,insoluble in glycerin | |
| C=CCCCCCCCCC=O | |
| 168.28 | |
| 168.28 | |
| Crystalline Powder or Crystals |
| 7°C | |
| 0.8450g/mL | |
| 76°C | |
| 96% min. (GC) | |
| C11H20O | |
| H2C=CH(CH2)8CHO | |
| 25 g | |
| 10-undecenal, undecylenic aldehyde, undecylenaldehyde, hendecenal, 10-undecen-1-al, 1-undecen-10-al, 10-undecylenealdehyde, 10-hendecenal, 10-undecenyl aldehyde, c-11 aldehyde, undecylenic | |
| OFHHDSQXFXLTKC-UHFFFAOYSA-N | |
| undec-10-enal | |
| 8187 | |
| 97% | |
| Undecylenic aldehyde, 97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Warning
EINECSNumber : 203-973-1
RTECSNumber : YQ2835000
TSCA : TSCA
RUO – Research Use Only