missing translation for 'onlineSavingsMsg'
Learn More

Undecylbenzene 98.0+%, TCI America™

Catalog No. U00105ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00105ML 5 mL
U001025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U00105ML Supplier TCI America Supplier No. U00105ML

Chemical Identifiers

CAS 6742-54-7
Molecular Formula C17H28
Molecular Weight (g/mol) 232.411
MDL Number MFCD00026549
InChI Key XBEADGFTLHRJRB-UHFFFAOYSA-N
Synonym 1-phenylundecane, benzene, undecyl, n-undecylbenzene, undecanylbenzene, undecane, 1-phenyl, alkyl c10-c13 benzene, unii-8c9lpo3q9t, benzene, c10-13-alkyl derivs., benzene, mono-c10-13-alkyl derivs., 8c9lpo3q9t
PubChem CID 23194
IUPAC Name undecylbenzene
SMILES CCCCCCCCCCCC1=CC=CC=C1

Specifications

Melting Point -5°C
Color Yellow
Boiling Point 160°C
Quantity 5 mL
Formula Weight 232.41
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Undecylbenzene
EINECSNumber (3)-0021
TSCA Yes