missing translation for 'onlineSavingsMsg'
Learn More

Undecanophenone 98.0+%, TCI America™

Catalog No. U0053500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U0053500G 500 g
U005325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U0053500G Supplier TCI America Supplier No. U0053500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4433-30-1
Molecular Formula C17H26O
Molecular Weight (g/mol) 246.394
MDL Number MFCD00051548
InChI Key LHJBFOGCFZHBAJ-UHFFFAOYSA-N
Synonym undecanophenone, n-undecanophenone, 1-phenylundecanone, 1-undecanone, 1-phenyl, decyl phenyl ketone, 1-phenyl-1-undecanone, 1-phenyl-undecan-1-one, 1-phenyl-1-undecanone #, acmc-209jx6
PubChem CID 78167
IUPAC Name 1-phenylundecan-1-one
SMILES CCCCCCCCCCC(=O)C1=CC=CC=C1

Specifications

Color White
Quantity 500 g
Formula Weight 246.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Undecanophenone