missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Undecanoic Acid 98.0+%, TCI America™
$52.19 - $246.05
Chemical Identifiers
| CAS | 112-37-8 |
|---|---|
| Molecular Formula | C11H22O2 |
| Molecular Weight (g/mol) | 186.295 |
| MDL Number | MFCD00002730 |
| InChI Key | ZDPHROOEEOARMN-UHFFFAOYSA-N |
| Synonym | hendecanoic acid, undecylic acid, n-undecanoic acid, n-undecoic acid, n-undecylic acid, undecoic acid, undecanoate, 1-decanecarboxylic acid, undekansaeure, unii-138on3iiqg |
| PubChem CID | 8180 |
| ChEBI | CHEBI:32368 |
| IUPAC Name | undecanoic acid |
| SMILES | CCCCCCCCCCC(=O)O |
Chemical Identifiers
| 112-37-8 | |
| 186.295 | |
| ZDPHROOEEOARMN-UHFFFAOYSA-N | |
| 8180 | |
| undecanoic acid |
| C11H22O2 | |
| MFCD00002730 | |
| hendecanoic acid, undecylic acid, n-undecanoic acid, n-undecoic acid, n-undecylic acid, undecoic acid, undecanoate, 1-decanecarboxylic acid, undekansaeure, unii-138on3iiqg | |
| CHEBI:32368 | |
| CCCCCCCCCCC(=O)O |
Specifications
| 112-37-8 | |
| 250°C | |
| MFCD00002730 | |
| hendecanoic acid, undecylic acid, n-undecanoic acid, n-undecoic acid, n-undecylic acid, undecoic acid, undecanoate, 1-decanecarboxylic acid, undekansaeure, unii-138on3iiqg | |
| CCCCCCCCCCC(=O)O | |
| 186.295 | |
| CHEBI:32368 | |
| ≥98.0% (GC,T) | |
| Undecanoic Acid |
| White | |
| C11H22O2 | |
| 25 g | |
| ZDPHROOEEOARMN-UHFFFAOYSA-N | |
| undecanoic acid | |
| 8180 | |
| 186.30 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (2)-0608
RTECSNumber : YQ2275000
TSCA : Yes