missing translation for 'onlineSavingsMsg'
Learn More

Undecanoic Acid 98.0+%, TCI America™

Catalog No. U000425G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U000425G 25 g
U0004250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U000425G Supplier TCI America Supplier No. U000425G

Chemical Identifiers

CAS 112-37-8
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.295
MDL Number MFCD00002730
InChI Key ZDPHROOEEOARMN-UHFFFAOYSA-N
Synonym hendecanoic acid, undecylic acid, n-undecanoic acid, n-undecoic acid, n-undecylic acid, undecoic acid, undecanoate, 1-decanecarboxylic acid, undekansaeure, unii-138on3iiqg
PubChem CID 8180
ChEBI CHEBI:32368
IUPAC Name undecanoic acid
SMILES CCCCCCCCCCC(=O)O

Specifications

Color White
Boiling Point 250°C
Quantity 25 g
Formula Weight 186.30
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material Undecanoic Acid
EINECSNumber (2)-0608
RTECSNumber YQ2275000
TSCA Yes