missing translation for 'onlineSavingsMsg'
Learn More

Undecane 99.0+%, TCI America™

Catalog No. U000225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U000225ML 25 mL
U0002500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U000225ML Supplier TCI America Supplier No. U000225ML

Chemical Identifiers

CAS 1120-21-4
Molecular Formula C11H24
Molecular Weight (g/mol) 156.31
MDL Number MFCD00008959
InChI Key RSJKGSCJYJTIGS-UHFFFAOYSA-N
Synonym n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und
PubChem CID 14257
ChEBI CHEBI:46342
IUPAC Name undecane
SMILES CCCCCCCCCCC

Specifications

Melting Point -26°C
Color Colorless
Boiling Point 195°C
Quantity 25 mL
UN Number 2330
Formula Weight 156.31
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Undecane
EINECSNumber (2)-0010
RTECSNumber YQ1525000
TSCA Yes