missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Undecafluoropentyl Iodide (stabilized with Copper chip) 98.0+%, TCI America™
$210.63 - $732.57
Chemical Identifiers
| CAS | 638-79-9 |
|---|---|
| Molecular Formula | C5F11I |
| Molecular Weight (g/mol) | 395.942 |
| MDL Number | MFCD08461629 |
| InChI Key | KCEJJSGJNCSQFI-UHFFFAOYSA-N |
| Synonym | Perfluoropentyl Iodide |
| PubChem CID | 69494 |
| IUPAC Name | 1,1,1,2,2,3,3,4,4,5,5-undecafluoro-5-iodopentane |
| SMILES | C(C(C(F)(F)F)(F)F)(C(C(F)(F)I)(F)F)(F)F |
Chemical Identifiers
| 638-79-9 | |
| 395.942 | |
| KCEJJSGJNCSQFI-UHFFFAOYSA-N | |
| 69494 | |
| C(C(C(F)(F)F)(F)F)(C(C(F)(F)I)(F)F)(F)F |
| C5F11I | |
| MFCD08461629 | |
| Perfluoropentyl Iodide | |
| 1,1,1,2,2,3,3,4,4,5,5-undecafluoro-5-iodopentane |
Specifications
| 638-79-9 | |
| Yellow | |
| C5F11I | |
| 5 g | |
| KCEJJSGJNCSQFI-UHFFFAOYSA-N | |
| 1,1,1,2,2,3,3,4,4,5,5-undecafluoro-5-iodopentane | |
| 69494 | |
| ≥98.0% (GC) | |
| Undecafluoropentyl Iodide (stabilized with Copper chip) |
| -50°C | |
| 94°C | |
| MFCD08461629 | |
| Perfluoropentyl Iodide | |
| C(C(C(F)(F)F)(F)F)(C(C(F)(F)I)(F)F)(F)F | |
| 395.942 | |
| 395.94 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0090
TSCA : Yes