missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Undecafluoropentyl Iodide (stabilized with Copper chip) 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | -50°C |
| Color | Yellow |
| Boiling Point | 94°C |
| Quantity | 5 g |
| Formula Weight | 395.94 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Undecafluoropentyl Iodide (stabilized with Copper chip) |
Safety and Handling
| EINECSNumber | (2)-0090 |
| TSCA | Yes |