missing translation for 'onlineSavingsMsg'
Learn More

Umbelliferone 98.0+%, TCI America™

Catalog No. p-7130429
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H02365G 5 g
H023625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H02365G Supplier TCI America Supplier No. H02365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 93-35-6
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00006878
InChI Key ORHBXUUXSCNDEV-UHFFFAOYSA-N
Synonym 7-hydroxycoumarin, umbelliferone, 7-hydroxy-2h-chromen-2-one, hydrangin, skimmetin, 7-hydroxycoumarine, 7-oxycoumarin, hydrangine, skimmetine, umbelliferon
PubChem CID 5281426
ChEBI CHEBI:27510
IUPAC Name 7-hydroxy-2H-chromen-2-one
SMILES OC1=CC=C2C=CC(=O)OC2=C1

Specifications

Melting Point 235°C
Quantity 5 g
Formula Weight 162.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Umbelliferone
EINECSNumber (5)-3651
RTECSNumber GN6820000
TSCA No