missing translation for 'onlineSavingsMsg'
Learn More

Ansell MICROFLEX™ UltraSense™ US-220 Nitrile Gloves

Catalog No. 296358564
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296358564 Small
296358563 X-Small
296358565 Medium
296358566 Large
296358567 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 296358564 Supplier Ansell Supplier No. US220S
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Powder free, comfortable nitrile examination gloves offer durability when worn for extended periods of time. Ideal for healthcare, life science and food processing applications.

  • High-quality nitrile offers reliable barrier protection against many hazards and infectious substances
  • Ultra soft nitrile provides the fit and feel of latex
  • Textured fingers provide secure grip in wet and dry environments
  • Non-sterile, ambidextrous, single-use
  • Length: 9.6 in. (24.4 cm)
  • Fingertip thickness: 4.3 mil. Palm thickness: 3.1 mil
  • Unique dark blue color

Specifications

Certifications/Compliance ASTM D6319, FDA21 CFR 177-2600
Certifications/Compliance (Europe) EN 374:2003, EN 420:2003 + A1:2009
Chemotherapeutic Rated No
Chlorinated Yes
Applications Industrial, Healthcare
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 4.3 mil
Finger Thickness (Metric) 0.11 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 3.1 mil
Palm Thickness (Metric) 0.08 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Small
Sterility Non-sterile
Thickness (English) 3.1 mil
Thickness (Metric) 0.07 mm
FDA 510 (k) Yes
Glove Cuff Style Beaded
Model US-220-S
Quantity 1000/Cs.
Series MICROFLEX™ UltraSense™ US-220
Tensile Strength 30 MPa
Length (English) 9.6 in.
Length (Metric) 24.38 cm
Show More Show Less