Learn More
MAPA™ Ultrane 524 ESD Glove
Catalog No. p-7221159
Change view
Click to view available options
Size:
10
11
6
7
8
9
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19046600 6
19046601 7
19046602 8
19046603 9
19046604 10
19046605 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19046600 Supplier MAPA™ Supplier No. 34524006
Only null left
Lightweight, PU coated, general purpose glove with electrostatic discharge properties and touchscreen capabilities.
  • ESD Properties
  • Touchscreen Capabilites
  • PU Coated
  • Lightweight
  • Comfort
  • Breathability
  • Silicon Free
  • Washable
Specifications
Coating Material Polyamide, Polyurethane
Color Gray
Cut Resistance Rating ANSI A1
Hand Specific
Latex Free No
Material Elastomer, PU
Size 6
Gender Unisex
Glove Cuff Style Continuous Knit
Puncture Rating ASTM F2878 LEVEL 0
Thumb Style Wing
Length (Metric) 27 cm
Show More Show Less