missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ UltraFit™ Nylon Medical Glove Liners

Catalog No. p-7230595
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
1437938 Medium
1437939 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1437938 Supplier Epredia™ Supplier No. 5100212001I
Only null left

Nylon glove liners are ideal for those who have sensitivities to latex, nitrile, and vinyl materials. A form fitting design and seamless fingertips ensure optimum dexterity and tactile sensitivity.

  • Inspected for medical use
  • Helps prevent irritation caused by latex, nylon, and vinyl gloves
  • Non-linting, ultra-thin nylon construction
  • Seamless fingertips
  • Form-fitting design
  • Washable

Specifications

Color White
Disposable Reusable
Hand Ambidextrous
Material Nylon
Size Medium
For Use With (Application) Medical Use
Glove Cuff Style Knit
Hem Color Orange
Inspected for Medical Use Yes