Learn More
Invitrogen™ UltraComp eBeads™ Plus Compensation Beads
Catalog No. p-7184690
Encompass
Change view
Click to view available options
No. of Tests:
100
25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Tests
01333341 25
01333342 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01333341 Supplier Invitrogen™ Supplier No. 01333341
Only null left
UltraComp eBeads Plus Compensation Beads are the second generation of UltraComp beads for use in flow cytometry.
  • Binds the spherical particles for use as a single-color compensation controls.
  • Binds with mouse, rat and hamster antibodies, but also bind with rabbit and recombinant human antibodies.
  • Designed for use in compensation with all fluorochromes excited by ultraviolet (355 nm) violet (405 nm), blue (488 nm), green (532 nm), yellow-green (561 nm), and red (633-640 nm) lasers.
  • Compensate in a manner similar to cells when used with Brilliant Violet 785/786-, Brilliant Violet 711-, Super Bright 780- and Super Bright 702-conjugated antibodies.
  • Multispecies compatibility—bind with mouse, rat, hamster, rabbit and recombinant human antibodies
  • Improved fluorophore compatibility
  • Each drop of beads contains two populations: a positive population that bind with any mouse, rat, hamster, rabbit or recombinant human antibody and a negative population that will not bind with antibody.
  • When a fluorochrome-conjugated antibody is added to the beads, both positive and negative populations are observed.
  • This bimodal distribution can be used for single-color compensation controls to accurately setup instrumentation for multicolor flow cytometry experiments.
Specifications
Product Line eBeads
No. of Tests 25
Quantity 25 tests
Product Type Compensation Beads
For Use With (Application) Flow Cytometry
RUO use only.